Få mere at vide
1-(3-Chlorophenyl)-4-(3-chloropropyl)piperazine Hydrochloride, TRC
Artikelnummer. 30363041
Skift visning
Mængde:
25 g
Artikelnummer. Mængde
30363041 25 g
1 options
Artikelnummer. 30363041 Leverandør Toronto Research Chemicals Leverandørnr. TRCC37772025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-(3-Chloropropyl)-4-(3-chlorophenyl)piperazine Hydrochloride
CAS 52605-52-4
Molekylær formel C13 H18 Cl2 N2 . Cl H
Mængde 25 g
Synonym 1-(3-Chloropropyl)-4-(3-chlorophenyl)piperazine Hydrochloride, Trazodone Hydrochloride Imp. F (BP) as Hydrochloride, Piperazine, 1-(3-chlorophenyl)-4-(3-chloropropyl)-, hydrochloride (1:1), Piperazine, 1-(3-chlorophenyl)-4-(3-chloropropyl)-, monohydrochloride (9CI), 1-(3-Chlorophenyl)-4-(3-chloropropyl)piperazine hydrochloride (1:1)
InChI formel InChI=1S/C13H18Cl2N2.ClH/c14-5-2-6-16-7-9-17(10-8-16)13-4-1-3-12(15)11-13;/h1,3-4,11H,2,5-10H2;1H
SMIL Cl.ClCCCN1CCN(CC1)c2cccc(Cl)c2
IUPAC navn 1-(3-chlorophenyl)-4-(3-chloropropyl)piperazine;hydrochloride
Molekylvægt (g/mol) 309.66
Formel vægt 308.0614