Få mere at vide
1-(3-Chloro-2-hydroxypropyl)-2-methyl-4-nitroimidazole, Mikromol™
Artikelnummer. 30052029
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052029 100 mg
30050452 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052029 Leverandør Mikromol Leverandørnr. MM0948.04
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-(3-Chloro-2-hydroxypropyl)-2-methyl-4-nitroimidazole
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-(3-Chloro-2-hydroxypropyl)-2-methyl-4-nitroimidazole
CAS 14419-11-5
Analyt- eller komponentnavne 1-(3-Chloro-2-hydroxypropyl)-2-methyl-4-nitroimidazole
Forsendelsestilstand Room Temperature
Molekylær formel C7 H10 Cl N3 O3
Certificeringer/compliance ISO 17025
Synonym 1-(3-Chloro-2-hydroxypropyl)-2-methyl-4-nitroimidazole
InChI formel InChI=1S/C7H10ClN3O3/c1-5-9-7(11(13)14)4-10(5)3-6(12)2-8/h4,6,12H,2-3H2,1H3
SMIL Cc1nc(cn1CC(O)CCl)[N+](=O)[O-]
IUPAC navn 1-chloro-3-(2-methyl-4-nitroimidazol-1-yl)propan-2-ol
Molekylvægt (g/mol) 219.63
Mængde 100 mg
Formel vægt 219.0411 g/mol