Få mere at vide
1,3-Bis-[4-(3-chlorophenyl)piperazin-1-yl]propane Dihydrochloride, Mikromol™
Artikelnummer. 30458139
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30458139 4 x 25 mg
17933436 25 mg
17993846 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30458139 Leverandør Mikromol Leverandørnr. MM0421.0800254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,3-Bis-[4-(3-chlorophenyl)piperazin-1-yl]propane Dihydrochloride, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,3-Bis-[4-(3-chlorophenyl)piperazin-1-yl]propane Dihydrochloride
CAS 2408971-27-5
Analyt- eller komponentnavne 1,3-Bis-[4-(3-chlorophenyl)piperazin-1-yl]propane Dihydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C23 H30 Cl2 N4 . 2 Cl H
Certificeringer/compliance ISO 17025
Synonym 1,3-Bis-[4-(3-chlorophenyl)piperazin-1-yl]propane Dihydrochloride, Trazodone Hydrochloride Imp. H (BP) as Dihydrochloride, Trazodone Hydrochloride Impurity H, Trazodone Impurity H
InChI formel InChI=1S/C23H30Cl2N4.2ClH/c24-20-4-1-6-22(18-20)28-14-10-26(11-15-28)8-3-9-27-12-16-29(17-13-27)23-7-2-5-21(25)19-23;;/h1-2,4-7,18-19H,3,8-17H2;2*1H
Løsningstype Neat
SMIL Cl.Cl.Clc1cccc(c1)N2CCN(CCCN3CCN(CC3)c4cccc(Cl)c4)CC2
IUPAC navn 1-(3-chlorophenyl)-4-[3-[4-(3-chlorophenyl)piperazin-1-yl]propyl]piperazine;dihydrochloride
Molekylvægt (g/mol) 506.34
Mængde 4 x 25 mg
Formel vægt 504.1381 g/mol