missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,3-Bis(3-boronophenyl)urea bis(pinacol) ester, 95%

Brand:  Alfa Aesar™ H64631.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469036

  • 2296.54 DKK / 1g


Estimated Shipment date
from Supplier 17-01-2019
Add to basket

Specifications

Chemical Identifiers

MFCD28100493
1,3-Bis[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]urea

Specifications

1,3-Bis(3-boronophenyl)urea bis(pinacol) ester
MFCD28100493
464.18
95%
1g
1,3-Bis[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]urea
Solid
Videos
SDS