Få mere at vide
1,3-Bis(2-acetyl-3-hydroxyphenoxy)-2-propanol, Mikromol™
Artikelnummer. 17972046
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17972046 100 mg
17920436 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17972046 Leverandør Mikromol Leverandørnr. MM0039.02
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,3-Bis(2-acetyl-3-hydroxyphenoxy)-2-propanol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,3-Bis(2-acetyl-3-hydroxyphenoxy)-2-propanol
CAS 16150-44-0
Analyt- eller komponentnavne 1,3-Bis(2-acetyl-3-hydroxyphenoxy)-2-propanol
Forsendelsestilstand Room Temperature
Molekylær formel C19 H20 O7
Certificeringer/compliance ISO 17025
Synonym 1,3-Bis(2-acetyl-3-hydroxyphenoxy)-2-propanol, Ethanone, 1,1'-[(2-hydroxy-1,3-propanediyl)bis[oxy(6-hydroxy-2,1-phenylene)]]bis- (9CI), Acetophenone, 2',2''-[(2-hydroxytrimethylene)dioxy]bis[6'-hydroxy- (8CI), 1,3-Bis(2-acetyl-3-hydroxyphenoxy)-2-hydroxypropane, Cromolyn Related Compound A
InChI formel InChI=1S/C19H20O7/c1-11(20)18-14(23)5-3-7-16(18)25-9-13(22)10-26-17-8-4-6-15(24)19(17)12(2)21/h3-8,13,22-24H,9-10H2,1-2H3
SMIL CC(=O)c1c(O)cccc1OCC(O)COc2cccc(O)c2C(=O)C
IUPAC navn 1-[2-[3-(2-acetyl-3-hydroxyphenoxy)-2-hydroxypropoxy]-6-hydroxyphenyl]ethanone
Molekylvægt (g/mol) 360.36
Mængde 100 mg
Formel vægt 360.1209 g/mol