Learn More
1,3-Benzothiazol-6-amine, 97+%, Maybridge™
Brand:  Maybridge CC05914EB
Additional Details : CAS Number : 533-30-2 Weight : 0.02500kg
Packaging Amber Glass Bottle
Quantity 25g
 View more versions of this product
Product Code. 10421474
  • 2668.37 DKK / 25g
Estimated Shipment
23-03-2021
Specifications
Chemical Identifiers
533-30-2
150.199
6-aminobenzothiazole, 6-benzothiazolamine, benzo d thiazol-6-amine, 6-amino-benzothiazole, benzothiazole, 6-amino, benzothiazol-6-amine, benzothiazol-6-ylamine, unii-9yfg4wuj1g, 9yfg4wuj1g, 1,3-benzothiazol-6-ylamine
1,3-benzothiazol-6-amine
C7H6N2S
FAYAYUOZWYJNBD-UHFFFAOYSA-N
68288
C1=CC2=C(C=C1N)SC=N2
Specifications
1, 3-Benzothiazol-6-amine
25g
97%
15, 420
FAYAYUOZWYJNBD-UHFFFAOYSA-N
1,3-benzothiazol-6-amine
68288
97%
Amber Glass Bottle
533-30-2
C7H6N2S
6-aminobenzothiazole, 6-benzothiazolamine, benzo d thiazol-6-amine, 6-amino-benzothiazole, benzothiazole, 6-amino, benzothiazol-6-amine, benzothiazol-6-ylamine, unii-9yfg4wuj1g, 9yfg4wuj1g, 1,3-benzothiazol-6-ylamine
C1=CC2=C(C=C1N)SC=N2
150.199
150.2
Videos
SDS Documents