Learn More
Alfa Aesar™ 1,3-Benzodithiolylium tetrafluoroborate, 98%
Brand:  Alfa Aesar™ H55403.06
Additional Details : CAS Number : 57842-27-0 Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15401638
  • 1215.37 DKK / 5g
In Stock 1
Add to basket
Specifications
Chemical Identifiers
57842-27-0
240.041
CUSWPJQKCZMDPY-UHFFFAOYSA-N
2735971
[B-](F)(F)(F)F.C1=CC=C2C(=C1)SC=[S+]2
C7H5BF4S2
MFCD00070613
1,3-benzodithiolylium tetrafluoroborate, benzo d 1,3 dithiol-1-ium tetrafluoroborate, 1,3-benzodithiolium tetrafluoroborate, 1,3-benzodithiol-3-ium tetrafluoroborate, 1,3-benzodithiol-2-ylium tetrafluoroborate, 1??,3-benzodithiol-1-ylium tetrafluoroborate, 2h-1,3-benzodithiol-2-ylium tetrafluoroborate, c7h5s2.bf4, 1,3-benzolithiolylium tetrafluoroborate, 1,3-benzodithiolylium tetrafluoroborate
1,3-benzodithiol-3-ium;tetrafluoroborate
Specifications
1,3-Benzodithiolylium tetrafluoroborate
150°C (decomposition)
57842-27-0
MFCD00070613
1,3-benzodithiolylium tetrafluoroborate, benzo d 1,3 dithiol-1-ium tetrafluoroborate, 1,3-benzodithiolium tetrafluoroborate, 1,3-benzodithiol-3-ium tetrafluoroborate, 1,3-benzodithiol-2-ylium tetrafluoroborate, 1??,3-benzodithiol-1-ylium tetrafluoroborate, 2h-1,3-benzodithiol-2-ylium tetrafluoroborate, c7h5s2.bf4, 1,3-benzolithiolylium tetrafluoroborate, 1,3-benzodithiolylium tetrafluoroborate
[B-](F)(F)(F)F.C1=CC=C2C(=C1)SC=[S+]2
240.041
240.04
98%
Brown to Gray
5g
C7H5BF4S2
4731453
CUSWPJQKCZMDPY-UHFFFAOYSA-N
1,3-benzodithiol-3-ium;tetrafluoroborate
2735971
Solid