missing translation for 'onlineSavingsMsg'
Få mere at vide

1,3,7-Trimethyluric Acid-d3, TRC

Artikelnummer. 30453983
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30453983 10 mg
30450373 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30453983 Leverandør Toronto Research Chemicals Supplier No. TRCT79800010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 188297-95-2
Molekylær formel C8 2H3 H7 N4 O3
Molekylvægt (g/mol) 213.21
IUPAC navn 1,3-dimethyl-7-(trideuteriomethyl)-9H-purine-2,6,8-trione
SMIL [2H]C([2H])([2H])N1C(=O)NC2=C1C(=O)N(C)C(=O)N2C

Specifications

Kemisk navn eller materiale 1,3,7-Trimethyluric Acid-d3
Mængde 10 mg
InChI formel InChI=1S/C8H10N4O3/c1-10-4-5(9-7(10)14)11(2)8(15)12(3)6(4)13/h1-3H3,(H,9,14)/i1D3
Formel vægt 213.0941
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.