Få mere at vide
1,3,5-Triphenylbenzene-d18, CDN
Artikelnummer. 30310981
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30310981 0.25 g -
30311269 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310981 Leverandør CDN Leverandørnr. CDND66160.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,3,5-Triphenylbenzene-D18
CAS 2838958-26-0
Molekylær formel C24 D18
Mængde 0.25 g
Synonym 5'-(Phenyl-d5)-1,1':3',1''-terphenyl-2,2',2'',3,3'',4,4',4'',5,5'',6,6',6''-d13, perdeuterated 1,3,5-Triphenylbenzene, 1,3,5-Triphenylbenzene-d18, 1,3,5-Triphenylbenzene-D18, 3,5-Di(phenyl-d5)-1,1'-biphenyl-2,2',3',4,4',5',6,6'-d8
InChI formel InChI=1 S/C24H18/c1-4-10-19(11-5-1)22-16-23(20-12-6-2-7-13-20)18-24(17-22)21-14-8-3-9-15-21/h1-18 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D,11 D,12 D,13 D,14 D,15 D,16 D,17 D,18 D
SMIL [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])c2c([2 H])c(c([2 H])c(c2[2 H])c3c([2 H])c([2 H])c([2 H])c([2 H])c3[2 H])c4c([2 H])c([2 H])c([2 H])c([2 H])c4[2 H]
IUPAC navn 1,2,3,4,5-pentadeuterio-6-[2,4,6-trideuterio-3,5-bis(2,3,4,5,6-pentadeuteriophenyl)phenyl]benzene
Molekylvægt (g/mol) 324.5106
Formel vægt 324.2538 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity