Få mere at vide
1,3,5-Trimethylbenzene-d12, CDN
Artikelnummer. 30309984
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30309984 1 g
30311370 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30309984 Leverandør CDN Leverandørnr. CDND29111G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,3,5-Trimethylbenzene D12
CAS 69441-16-3
Molekylær formel C9 D12
Mængde 1 g
Synonym 1,3,5-Trimethylbenzene D12, Mesitylene-d12 (6 CI), 2,4,6-Tri(methyl-d3)benzene-1,3,5-d3, 1,3,5-Trideuterio-2,4,6-tris(trideuteriomethyl)benzene, 2,4,6-Trideutero-1,3,5-tris(trideuteromethyl)benzene, Perdeuterated 1,3,5-trimethylbenzene, Perdeuteromesitylene
InChI formel InChI=1 S/C9H12/c1-7-4-8(2)6-9(3)5-7/h4-6 H,1-3H3/i1D3,2D3,3D3,4 D,5 D,6 D
SMIL [2 H]c1c(c([2 H])c(c([2 H])c1C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,3,5-trideuterio-2,4,6-tris(trideuteriomethyl)benzene
Molekylvægt (g/mol) 132.266
Formel vægt 132.169 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity