Få mere at vide
1,3,5-Trimethylbenzene-2,4,6-d3, TRC
Artikelnummer. 30472270
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30472270 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30472270 Leverandør Toronto Research Chemicals Leverandørnr. TRCM25839110MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,3,5-Trimethylbenzene-2,4,6-d3
CAS 38574-14-0
Molekylær formel C9 D3 H9
Mængde 10 mg
Synonym Benzene-1,3,5-d3, 2,4,6-trimethyl- (9CI, ACI), 2,4,6-Trimethylbenzene-1,3,5-d3 (ACI), 2,4,6-Trideutero-1,3,5-trimethylbenzene, Mesitylene-d3, 1,3,5-Trimethylbenzene-d3, 1,3,5-Trimethylbenzene-2,4,6-D3
InChI formel InChI=1S/C9H12/c1-7-4-8(2)6-9(3)5-7/h4-6H,1-3H3/i4D,5D,6D
SMIL [2H]c1c(C)c([2H])c(C)c([2H])c1C
IUPAC navn 1,3,5-trideuterio-2,4,6-trimethylbenzene
Molekylvægt (g/mol) 123.2101
Formel vægt 123.1127