Få mere at vide
enQuireBio™ Recombinant other Protein-A/G/L-Cys Protein
Artikelnummer. p-7151049
Skift visning
Mængde:
1 mg
10 mg
100 mg
Pakningsstørrelse:
100mg
10mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
15922709 1 mg 1mg
15912709 10 mg 10mg
15902709 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 15922709 Leverandør enQuireBio™ Leverandørnr. QP131661mg
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
A cDNA sequence encoding the Protein-A/G/L-Cys was constructed and used to recombinantly synthesize the protein.
Tekniske data
Navn Protein-A/G/L-Cys Protein
Mængde 1 mg
Regulatorisk status RUO
Endotoksin koncentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Produkttype Recombinant Protein
Krydsreaktivitet Other
Arter E. coli
Protein tag Untagged
Sekvens NAAQHDEAQQ NAFYQVLNMP NLNADQRNGF IQSLKDDPSQ SANVLGEAQK LNDSQAPKAD AQQNNFNKDQ QSAFYEILNM PNLNEAQRNG FIQSLKDDPS QSTNVLGEAK KLNESQAPKA DNNFNKEQQN AFYEILNMPN LNEEQRNGFI QSLKDDPSQS ANLLSEAKKL NESQAPKADN KFNKEQQNAF YEILHLPNLN EEQRNGFIQS LKDDPSQSAN LLAEAKKLND AQAPKADNKF NKEQQNAFYE ILHLPNLTEE QRNGFIQSLK DDPSVSKEIL AEAKKLNDAQ APKEEDSLEG SGSGTYKLIL NGKTLKGETT TEAVDAATAE KVFKQYANDN GVDGEWTYDD ATKTFTVTEK PEVIDASELT PAVTTYKLVI NGKTLKGETT TKAVDAETAE KAFKQYANDN GVDGVWTYDD ATKTFTVTEE PRARPGSGSG KEETPETPET DSEEEVTIKA NLIFANGSTQ TAEFKGTFEK ATSEAYAYAD TLKKDNGEYT VDVADKGYTL NIKFAGKEKT PEEPKEEVTI KANLIYADGK TQTAEFKGTF EEATAEAYRY ADALKKDNGE YTVDVADKGY TLNIKFAGKE KTPEEPKEEV TIKANLIYAD GKTQTAEFKG TFEEATAEAY RYADLLAKEN GKYTVDVADK GYTLNIKFAG KEKTPEEPKE EVTIKANLIY ADGKTQTAEF KGTFAEATAE AYRYADLLAK ENGKYTADLE DGGYTINIRF AGKKVDEKPE EKEQVTIKEN IYFEDGTVQT ATFKGTFAEA TAEAYRYADL LSKEHGKYTA DLEDGGYTIN IRFAGC
Buffer Protein- A/G/L was lyophilized without any additives.
Renhed eller kvalitetsklasse Greater than 95.0% as determined by: (a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE.