Få mere at vide
1,3,5-Trichloro-2-methylbenzene, TRC
Artikelnummer. 17942005
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17942005 50 mg
17930985 25 mg
17902195 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17942005 Leverandør Toronto Research Chemicals Leverandørnr. TRCT77436350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 23749-65-7
Molekylær formel C7 H5 Cl3
Molekylvægt (g/mol) 195.47
Synonym 1,3,5-Trichloro-2-methylbenzene (ACI), Toluene, 2,4,6-trichloro- (8CI), 2,4,6-Trichlorotoluene, 1,3,5-Trichloro-2-methylbenzene (ACI), Toluene, 2,4,6-trichloro- (8CI), 2,4,6-Trichlorotoluene, 2,4,6-Trichloro-toluene;
IUPAC navn 1,3,5-trichloro-2-methylbenzene
SMIL Cc1c(Cl)cc(Cl)cc1Cl
Tekniske data
Kemisk navn eller materiale 1,3,5-Trichloro-2-methylbenzene
Mængde 50 mg
Renhedsgrad noter GC
InChI formel InChI=1S/C7H5Cl3/c1-4-6(9)2-5(8)3-7(4)10/h2-3H,1H3
Formel vægt 193.946
Procent renhed >95%