Learn More
1-(3,5-Dichlorophenyl)ethanone, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 14401-72-0 |
| Molecular Formula | C8H6Cl2O |
| MDL Number | MFCD00045189 |
| Quantity | 1 g |
| SMILES | CC(C1=CC(Cl)=CC(Cl)=C1)=O |
| Molecular Weight (g/mol) | 189.04 |
| Percent Purity | 97% |
| Chemical Name or Material | 1-(3,5-Dichlorophenyl)ethanone |