Få mere at vide
1,3,4,5-Tetrahydro-7,8-dimethoxy-3-[3-(methylamino)propyl]-2H-3-benzazepin-2-one, TRC
Artikelnummer. 30415197
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30415197 50 mg
30420186 5 mg
30420797 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30415197 Leverandør Toronto Research Chemicals Leverandørnr. TRCT29407050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 85175-77-5
Molekylær formel C16H24N2O3
Molekylvægt (g/mol) 292.37
Synonym 1,3,4,5-Tetrahydro-7,8-dimethoxy-3-[3-(methylamino)propyl]-2H-3-benzazepin-2-one
IUPAC navn 7,8-dimethoxy-3-[3-(methylamino)propyl]-2,5-dihydro-1H-3-benzazepin-4-one
SMIL CNCCCN1CCc2cc(OC)c(OC)cc2CC1=O
Tekniske data
Kemisk navn eller materiale 1,3,4,5-Tetrahydro-7,8-dimethoxy-3-[3-(methylamino)propyl]-2H-3-benzazepin-2-one
Mængde 50 mg
InChI formel InChI=1S/C16H24N2O3/c1-17-6-4-7-18-8-5-12-9-14(20-2)15(21-3)10-13(12)11-16(18)19/h9-10,17H,4-8,11H2,1-3H3
Formel vægt 292.1787