missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,3-Trimethyl-2-methyleneindoline, 98.5%, Thermo Scientific™
Click to view available options
Quantity:
250 g
Packaging:
Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
CAS Min % | 98.0 |
CAS Max % | 100.0 |
Melting Point | -8.0°C |
Color | Red |
Density | 0.9860g/mL |
Boiling Point | 106.0°C to 110.0°C (5.0 Mbar) |
Flash Point | 94°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Show More |
Safety and Handling
Product Identifier
- 1,3,3-Trimethyl-2-methyleneindoline
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Skin corrosion/irritation Category 2
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- H302-Harmful if swallowed.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'