Få mere at vide
1-[(2RS)-2-Benzyloxy-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate, Mikromol™
Beskrivelse
1-[(2RS)-2-Benzyloxy-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 1-[(2RS)-2-Benzyloxy-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate |
| CAS | 203264-09-9 |
| Analyt- eller komponentnavne | 1-[(2RS)-2-Benzyloxy-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C18 H16 Cl2 N2 O . H N O3 |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 1-[(2RS)-2-Benzyloxy-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate, Miconazole Nitrate Imp. H (EP) as Nitrate, Miconazole Impurity H as Nitrate, Miconazole Nitrate Impurity H as Nitrate, Miconazole Impurity H as Nitrate |
| InChI formel | InChI=1S/C18H16Cl2N2O.HNO3/c19-15-6-7-16(17(20)10-15)18(11-22-9-8-21-13-22)23-12-14-4-2-1-3-5-14;2-1(3)4/h1-10,13,18H,11-12H2;(H,2,3,4) |
| SMIL | O[N+](=O)[O-].Clc1ccc(C(Cn2ccnc2)OCc3ccccc3)c(Cl)c1 |
| IUPAC navn | 1-[2-(2,4-dichlorophenyl)-2-phenylmethoxyethyl]imidazole;nitric acid |
| Vis mere |