Få mere at vide
1-[(2RS)-2,3-Dihydro-1,4-benzodioxin-2-ylcarbonyl]piperazine Hydrochloride, Mikromol™
Beskrivelse
1-[(2RS)-2,3-Dihydro-1,4-benzodioxin-2-ylcarbonyl]piperazine Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 1-[(2RS)-2,3-Dihydro-1,4-benzodioxin-2-ylcarbonyl]piperazine Hydrochloride |
| CAS | 70918-74-0 |
| Analyt- eller komponentnavne | 1-[(2RS)-2,3-Dihydro-1,4-benzodioxin-2-ylcarbonyl]piperazine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C13 H16 N2 O3 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Doxazosin Mesilate Imp. B (EP), Doxazosin USP Related Compound A, Doxazosin USP RC A, 1-[(2RS)-2,3-Dihydro-1,4-benzodioxin-2-ylcarbonyl]piperazine Hydrochloride, Doxazosin Mesilate Imp. B (EP) as Hydrochloride, Doxazosin Related Compound A as Hydrochloride, Doxazosin Mesilate Impurity B as Hydrochloride, Doxazosin Impurity B as Hydrochloride |
| InChI formel | InChI=1S/C13H16N2O3.ClH/c16-13(15-7-5-14-6-8-15)12-9-17-10-3-1-2-4-11(10)18-12;/h1-4,12,14H,5-9H2;1H |
| SMIL | Cl.O=C(C1COc2ccccc2O1)N3CCNCC3 |
| IUPAC navn | 2,3-dihydro-1,4-benzodioxin-3-yl(piperazin-1-yl)methanone;hydrochloride |
| Vis mere |