Få mere at vide
1-[(2RS)-2-[(2-Chlorobenzyl)oxy]-2-(2,4-dichloro-phenyl)ethyl]-1H-imidazole Nitrate, Mikromol™
Artikelnummer. 30051159
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051159 100 mg
30049274 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051159 Leverandør Mikromol Leverandørnr. MM0281.09
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-[(2RS)-2-[(2-Chlorobenzyl)oxy]-2-(2,4-dichloro-phenyl)ethyl]-1H-imidazole Nitrate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-[(2RS)-2-[(2-Chlorobenzyl)oxy]-2-(2,4-dichloro-phenyl)ethyl]-1H-imidazole Nitrate
CAS 24169-00-4
Analyt- eller komponentnavne 1-[(2RS)-2-[(2-Chlorobenzyl)oxy]-2-(2,4-dichloro-phenyl)ethyl]-1H-imidazole Nitrate
Forsendelsestilstand Room Temperature
Molekylær formel C18 H15 Cl3 N2 O . H N O3
Certificeringer/compliance ISO 17025
Synonym Miconazole Nitrate Imp. I (EP), Miconazole USP Related Compound I, Miconazole USP RC I, 1-[(2RS)-2-[(2-Chlorobenzyl)oxy]-2-(2,4-dichloro-phenyl)ethyl]-1H-imidazole Nitrate, Miconazole Nitrate Imp. I (EP) as Nitrate, Miconazole Related Compound I as Nitrate, Miconazole Impurity I as Nitrate, Miconazole Nitrate Impurity I as Nitrate, Miconazole Impurity I as Nitrate
InChI formel InChI=1S/C18H15Cl3N2O.HNO3/c19-14-5-6-15(17(21)9-14)18(10-23-8-7-22-12-23)24-11-13-3-1-2-4-16(13)20;2-1(3)4/h1-9,12,18H,10-11H2;(H,2,3,4)
SMIL O[N+](=O)[O-].Clc1ccc(C(Cn2ccnc2)OCc3ccccc3Cl)c(Cl)c1
IUPAC navn 1-[2-[(2-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole;nitric acid
Molekylvægt (g/mol) 444.7
Mængde 100 mg
Formel vægt 443.0206 g/mol