Få mere at vide
1-[(2RS)-2-[(2-Chlorobenzyl)oxy]-2-(2,4-dichloro-phenyl)ethyl]-1H-imidazole Nitrate, Mikromol™
Beskrivelse
1-[(2RS)-2-[(2-Chlorobenzyl)oxy]-2-(2,4-dichloro-phenyl)ethyl]-1H-imidazole Nitrate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 1-[(2RS)-2-[(2-Chlorobenzyl)oxy]-2-(2,4-dichloro-phenyl)ethyl]-1H-imidazole Nitrate |
| CAS | 24169-00-4 |
| Analyt- eller komponentnavne | 1-[(2RS)-2-[(2-Chlorobenzyl)oxy]-2-(2,4-dichloro-phenyl)ethyl]-1H-imidazole Nitrate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C18 H15 Cl3 N2 O . H N O3 |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Miconazole Nitrate Imp. I (EP), Miconazole USP Related Compound I, Miconazole USP RC I, 1-[(2RS)-2-[(2-Chlorobenzyl)oxy]-2-(2,4-dichloro-phenyl)ethyl]-1H-imidazole Nitrate, Miconazole Nitrate Imp. I (EP) as Nitrate, Miconazole Related Compound I as Nitrate, Miconazole Impurity I as Nitrate, Miconazole Nitrate Impurity I as Nitrate, Miconazole Impurity I as Nitrate |
| InChI formel | InChI=1S/C18H15Cl3N2O.HNO3/c19-14-5-6-15(17(21)9-14)18(10-23-8-7-22-12-23)24-11-13-3-1-2-4-16(13)20;2-1(3)4/h1-9,12,18H,10-11H2;(H,2,3,4) |
| SMIL | O[N+](=O)[O-].Clc1ccc(C(Cn2ccnc2)OCc3ccccc3Cl)c(Cl)c1 |
| IUPAC navn | 1-[2-[(2-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole;nitric acid |
| Vis mere |