Få mere at vide
1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[[4-(benzenesulfinyl)benzyl]oxy]ethyl]-1H-imidazole Nitrate (Fenticonazole Sulfoxide Nitrate), Mikromol™
Beskrivelse
1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[[4-(benzenesulfinyl)benzyl]oxy]ethyl]-1H-imidazole Nitrate (Fenticonazole Sulfoxide Nitrate)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Fenticonazole Sulfoxide Nitrate |
| CAS | 80639-95-8 |
| Analyt- eller komponentnavne | Fenticonazole Sulfoxide Nitrate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C24 H20 Cl2 N2 O2 S . H N O3 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 1-[2-(2,4-Dichlorophenyl)-2-[[4-(phenylsulfinyl)phenyl]methoxy]ethyl]-1H-imidazole Nitrate, Fenticonazole Sulfoxide Nitrate, 1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[[4-(benzenesulfinyl)benzyl]oxy]ethyl]-1H-imidazole Nitrate, Fenticonazole Nitrate Impurity B as Nitrate, Fenticonazole Impurity B as Nitrate |
| InChI formel | InChI=1S/C24H20Cl2N2O2S.HNO3/c25-19-8-11-22(23(26)14-19)24(15-28-13-12-27-17-28)30-16-18-6-9-21(10-7-18)31(29)20-4-2-1-3-5-20;2-1(3)4/h1-14,17,24H,15-16H2;(H,2,3,4) |
| SMIL | O[N+](=O)[O-].Clc1ccc(C(Cn2ccnc2)OCc3ccc(cc3)S(=O)c4ccccc4)c(Cl)c1 |
| IUPAC navn | 1-[2-[[4-(benzenesulfinyl)phenyl]methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole;nitric acid |
| Vis mere |