Få mere at vide
1-((2R)-2-Amino-2-cyclohexylacetyl)-N-(4’-cyanobenzyl)-2-L-azetidinecarboxamide-d11, TRC
Artikelnummer. 30310836
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30310836 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310836 Leverandør Toronto Research Chemicals Leverandørnr. TRCA60366210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-((2R)-2-Amino-2-cyclohexylacetyl)-N-(4'-cyanobenzyl)-2-L-azetidinecarboxamide-d11
CAS 1356930-51-2
Molekylær formel C20 H15 D11 N4 O2
Mængde 10 mg
Synonym (2S)-1-((2R)-2-Amino-2-cyclohexylacetyl)-N-[[4-cyanophenyl]methyl]-2-azetidinecarboxamide-d11
InChI formel InChI=1S/C20H26N4O2/c21-12-14-6-8-15(9-7-14)13-23-19(25)17-10-11-24(17)20(26)18(22)16-4-2-1-3-5-16/h6-9,16-18H,1-5,10-11,13,22H2,(H,23,25)/t17-,18+/m0/s1/i1D2,2D2,3D2,4D2,5D2,16D
SMIL [2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])([C@@H](N)C(=O)N2CC[C@H]2C(=O)NCc3ccc(cc3)C#N)C([2H])([2H])C1([2H])[2H]
IUPAC navn (2S)-1-[(2R)-2-amino-2-(1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexyl)acetyl]-N-[(4-cyanophenyl)methyl]azetidine-2-carboxamide
Molekylvægt (g/mol) 365.51
Formel vægt 365.2746