missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Thien-2-ylphenyl)methanamine hydrochloride, ≥95%, Thermo Scientific™

Catalog Number p-3176130
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
863991-95-1
C11H12ClNS
225.73
MFCD07772821
XLJIHGHEJAGJJC-UHFFFAOYSA-N
1-2-thien-2-ylphenyl methanamine hydrochloride, 1-2-thien-2-ylphenyl methylamine hydrochloride, 2-thiophen-2-yl phenyl methanamine hydrochloride, 1-2-thien-2-ylphenyl methanaminehydrochloride, 1-2-thiophen-2-yl phenyl methanamine hydrochloride, c11h11ns.hcl, 2-thiophen-2-yl-benzylamine hydrochloride, 2-2-thienyl phenyl methylamine, chloride, 2-thien-2-yl phenyl methylamine hydrochloride
18525724
1-[2-(thiophen-2-yl)phenyl]methanamine hydrochloride
Cl.NCC1=CC=CC=C1C1=CC=CS1
This item is not returnable. View return policy
863991-95-1
C11H12ClNS
225.73
MFCD07772821
XLJIHGHEJAGJJC-UHFFFAOYSA-N
1-2-thien-2-ylphenyl methanamine hydrochloride, 1-2-thien-2-ylphenyl methylamine hydrochloride, 2-thiophen-2-yl phenyl methanamine hydrochloride, 1-2-thien-2-ylphenyl methanaminehydrochloride, 1-2-thiophen-2-yl phenyl methanamine hydrochloride, c11h11ns.hcl, 2-thiophen-2-yl-benzylamine hydrochloride, 2-2-thienyl phenyl methylamine, chloride, 2-thien-2-yl phenyl methylamine hydrochloride
18525724
1-[2-(thiophen-2-yl)phenyl]methanamine hydrochloride
Cl.NCC1=CC=CC=C1C1=CC=CS1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 225.74
Percent Purity 95%
Chemical Name or Material 1-(2-Thien-2-ylphenyl)methanamine hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'