Få mere at vide
(+/-)-1,2-Propylene-d6 Oxide (Stabilized with hydroquinone), TRC
Artikelnummer. 30412212
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30412212 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30412212 Leverandør Toronto Research Chemicals Leverandørnr. TRCP83524250MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,2-Propylene oxide-d6
CAS 202468-69-7
Molekylær formel C3 2H6 O
Mængde 50 mg
Synonym Oxirane-d3','methyl-d3- (9CI), (±)-1,2-Propylene-d6 oxide, Propylene Oxide-D6, Methyl-d3-oxirane-d3
InChI formel InChI=1S/C3H6O/c1-3-2-4-3/h3H,2H2,1H3/i1D3,2D2,3D
SMIL [2H]C([2H])([2H])C1([2H])OC1([2H])[2H]
IUPAC navn 2,2,3-trideuterio-3-(trideuteriomethyl)oxirane
Molekylvægt (g/mol) 64.12
Formel vægt 64.0795