Få mere at vide
(+/-)-1,2-Propylene-d6 Oxide (Stabilized with hydroquinone), TRC
Artikelnummer. 30419524
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30419524 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30419524 Leverandør Toronto Research Chemicals Leverandørnr. TRCP835242500MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,2-Propylene oxide-d6
CAS 202468-69-7
Molekylær formel C3 2H6 O
Mængde 500 mg
Synonym Oxirane-d3','methyl-d3- (9CI), (±)-1,2-Propylene-d6 oxide, Propylene Oxide-D6, Methyl-d3-oxirane-d3
InChI formel InChI=1S/C3H6O/c1-3-2-4-3/h3H,2H2,1H3/i1D3,2D2,3D
SMIL [2H]C([2H])([2H])C1([2H])OC1([2H])[2H]
IUPAC navn 2,2,3-trideuterio-3-(trideuteriomethyl)oxirane
Molekylvægt (g/mol) 64.12
Formel vægt 64.0795