Få mere at vide
1,2-Propylene-d6 Carbonate, TRC
Artikelnummer. 30422254
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30422254 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30422254 Leverandør Toronto Research Chemicals Leverandørnr. TRCP8367521G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,2-Propylene-d6 Carbonate
CAS 202480-74-8
Molekylær formel C4 2H6 O3
Mængde 1 g
Synonym 1,3-Dioxolan-2-one-4,4,5-d3','5-(methyl-d3)- (9CI), Propylene-d6 carbonate, 1,2-Propylene-d6 carbonate, 1,2-Propylene-D6 carbonate, 5-(Methyl-d3)-1,3-dioxolan-2-one-4,4,5-d3
InChI formel InChI=1S/C4H6O3/c1-3-2-6-4(5)7-3/h3H,2H2,1H3/i1D3,2D2,3D
SMIL [2H]C([2H])([2H])C1([2H])OC(=O)OC1([2H])[2H]
IUPAC navn 4,4,5-trideuterio-5-(trideuteriomethyl)-1,3-dioxolan-2-one
Molekylvægt (g/mol) 108.13
Formel vægt 108.0694