Få mere at vide
(±)-1,2-Propanediol-(OD)2, CDN
Artikelnummer. 30311656
Skift visning
Mængde:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30311656 10 g
30314721 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311656 Leverandør CDN Leverandørnr. CDND038610G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,2-Propanediol D2 (Di-O-D2)
CAS 58161-11-8
Molekylær formel C3 D2 H6 O2
Mængde 10 g
Synonym 1,2-Propanediol D2 (Di-O-D2), 1,2-Propanediol-d2 (7 CI,9 CI)
InChI formel InChI=1 S/C3H8O2/c1-3(5)2-4/h3-5 H,2H2,1H3/i4D,5 D
SMIL [2 H]OCC(C)O[2 H]
IUPAC navn 1,2-dideuteriooxypropane
Molekylvægt (g/mol) 78.107
Formel vægt 78.065 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity