Få mere at vide
1,2-Propanediol-d6, TRC
Artikelnummer. 30417198
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30417198 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30417198 Leverandør Toronto Research Chemicals Leverandørnr. TRCP760312250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,2-Propanediol D6
CAS 52910-80-2
Molekylær formel C3 D6 H2 O2
Mængde 250 mg
Synonym 1,2-Propanediol D6, 1,2-Propane-d6-diol, 1,2-Propane-1,1,2,3,3,3-d6-diol (9CI)
InChI formel InChI=1S/C3H8O2/c1-3(5)2-4/h3-5H,2H2,1H3/i1D3,2D2,3D
SMIL [2H]C([2H])([2H])C([2H])(O)C([2H])([2H])O
IUPAC navn 1,1,2,3,3,3-hexadeuteriopropane-1,2-diol
Molekylvægt (g/mol) 82.1314
Formel vægt 82.0901