missing translation for 'onlineSavingsMsg'
Få mere at vide

1,2-Propanediol-d3, TRC

Artikelnummer. 30420704
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30420704 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30420704 Leverandør Toronto Research Chemicals Leverandørnr. TRCP76031150MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 1,2-Propanediol-d3
CAS 70195-21-0
Molekylær formel C3 H5 D3 O2
Mængde 50 mg
Synonym (RS)-1,2-Propanediol-d3, (±)-1,2-Propanediol-d3, (±)-Propylene glycol-d3, 1,2-(RS)-Propanediol-d3, 1,2-Dihydroxypropane-d3, 1,2-Propylene glycol-d3, 1000PG-d3, 2,3-Propanedio-d3l, 2-Hydroxypropanol-d3, Adeka PG-d3, Adeka Propylene Glycol PG-P-d3, DC 403-d3, DL-1,2-Propanediol-d3, Dowfrost-d3, Isopropylene Glycol-d3, Kilfrost ABC-S-d3, Kollisolv PG-d3, Methylethyl Glycol-d3, Methylethylene Glycol-d3, Monopropylene Glycol-d3, NSC 69860-d3, Nybrine NFP-d3, PG 12-d3, PG-T-d3, PG-T (Glycol)-d3, ProGlyc 55-d3, Propylene Glycol-d3, Propyless-d3, Safewing MP-I 1938-d3, Safewing MP-IV 2001-d3, Sirlene-d3, Solar Winter Ban-d3, Solargard P-d3, Ucar 35-d3, dl-Propylene Glycol-d3, α-Propylene Glycol-d3
InChI formel InChI=1S/C3H8O2/c1-3(5)2-4/h3-5H,2H2,1H3/i2D2,3D
SMIL [2H]C([2H])(O)C([2H])(C)O
IUPAC navn 1,1,2-trideuteriopropane-1,2-diol
Molekylvægt (g/mol) 79.11
Formel vægt 79.0713
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.