Få mere at vide
(±)-1,2-Propane-d6-diol, CDN
Artikelnummer. 30322273
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30322273 1 g -
30319109 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322273 Leverandør CDN Leverandørnr. CDND15071G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,2-Propanediol D6
CAS 52910-80-2
Molekylær formel C3 D6 H2 O2
Mængde 1 g
Synonym 1,2-Propane-d6-diol, 1,2-Propane-1,1,2,3,3,3-d6-diol (9 CI)
InChI formel InChI=1 S/C3H8O2/c1-3(5)2-4/h3-5 H,2H2,1H3/i1D3,2D2,3 D
SMIL [2 H]C([2 H])([2 H])C([2 H])(O)C([2 H])([2 H])O
IUPAC navn 1,1,2,3,3,3-hexadeuteriopropane-1,2-diol
Molekylvægt (g/mol) 82.1314
Formel vægt 82.0901 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity