Få mere at vide
1,2-Phenylenediamine-d8, TRC
Artikelnummer. 30428563
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30428563 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30428563 Leverandør Toronto Research Chemicals Leverandørnr. TRCP319843250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,2-Phenylenediamine-d8
CAS 1219798-78-3
Molekylær formel C6D8N2
Mængde 250 mg
Synonym 1,2-Benzenediamine-d8, o-Phenylenediamine-d8, 1,2-Diaminobenzene-d8, 2-Aminoaniline-d8, C.I. 76010-d8, C.I. Oxidation Base 16-d8, IK 3, NSC 5354-d8, Orthamine-d8, o-Aminoaniline-d8, o-Aminophenylamine-d8, o-Benzenediamine-d8, o-Diaminobenzene-d8
InChI formel InChI=1S/C6H8N2/c7-5-3-1-2-4-6(5)8/h1-4H,7-8H2/i1D,2D,3D,4D/hD4
SMIL [2H]C1=C(N([2H])[2H])C(N([2H])[2H])=C([2H])C([2H])=C1[2H]
IUPAC navn benzene-3,4,5,6-d4-1,2-diamine-N1,N1,N2,N2-d4
Molekylvægt (g/mol) 116.19
Formel vægt 116.12