missing translation for 'onlineSavingsMsg'
Få mere at vide

1,2-Phenylenediamine-d4, TRC

Artikelnummer. 30420222
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30420222 25 mg
30415030 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30420222 Supplier Toronto Research Chemicals Supplier No. TRCP31984225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 291765-93-0
Molekylær formel C6 D4 H4 N2
Molekylvægt (g/mol) 112.17
Synonym 1,2-Benzenediamine-d4, 1,2-Benzene-d4-diamine, o-Phenylenediamine-d4, 1,2-Diaminobenzene-d4, 1,2-Benzene-3,4,5,6-d4-diamine, 3,4,5,6-Tetradeuteriobenzene-1,2-diamine, 2-Aminoaniline-d4, C.I. 76010-d4, C.I. Oxidation Base 16-d4, IK 3, NSC 5354-d4, Orthamine-d4, o-Aminoaniline-d4, o-Aminophenylamine-d4, o-Benzenediamine-d4, o-Diaminobenzene-d4, o-Phenylenediamine-d4, 1,2-Diaminobenzene-3,4,5,6-d4, Benzene-1,2-diamine-D4
IUPAC navn 3,4,5,6-tetradeuteriobenzene-1,2-diamine
SMIL [2H]c1c([2H])c([2H])c(N)c(N)c1[2H]

Specifications

Kemisk navn eller materiale 1,2-Phenylenediamine-d4
Mængde 25 mg
InChI formel InChI=1S/C6H8N2/c7-5-3-1-2-4-6(5)8/h1-4H,7-8H2/i1D,2D,3D,4D
Formel vægt 112.0939
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.