Få mere at vide
1-(2-Phenylacetyl)-L-prolylglycine Ethyl Ester, TRC
Artikelnummer. 30426225
Skift visning
Mængde:
1 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30426225 100 mg
30420742 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30426225 Leverandør Toronto Research Chemicals Leverandørnr. TRCP306600100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 157115-85-0
Molekylær formel C17 H22 N2 O4
Molekylvægt (g/mol) 318.37
Synonym Glycine, 1-(2-phenylacetyl)-L-prolyl-, ethyl ester, Glycine, 1-(phenylacetyl)-L-prolyl-, ethyl ester (9CI), Glycine, N-[1-(phenylacetyl)-L-prolyl]-, ethyl ester, 1-(2-Phenylacetyl)-L-prolylglycine ethyl ester, GVS 111, Noopept, Omberacetam, SGS 111
IUPAC navn ethyl 2-[[(2S)-1-(2-phenylacetyl)pyrrolidine-2-carbonyl]amino]acetate
SMIL CCOC(=O)CNC(=O)[C@@H]1CCCN1C(=O)Cc2ccccc2
Tekniske data
Kemisk navn eller materiale N-Phenylacetyl-L-prolylglycine ethyl ester
Mængde 100 mg
InChI formel InChI=1S/C17H22N2O4/c1-2-23-16(21)12-18-17(22)14-9-6-10-19(14)15(20)11-13-7-4-3-5-8-13/h3-5,7-8,14H,2,6,9-12H2,1H3,(H,18,22)/t14-/m0/s1
Formel vægt 318.158