missing translation for 'onlineSavingsMsg'
Få mere at vide

1,2-O-Isopropylidene-6-methyl-sulfonyl-Alpha-D-glucofuranose, TRC

Artikelnummer. 30403110
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30403110 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30403110 Leverandør Toronto Research Chemicals Leverandørnr. TRCI82527510G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 1,2-O-Isopropylidene-6-methyl-sulfonyl-a-D-glucofuranose
CAS 33557-25-4
Molekylær formel C10 H18 O8 S
Mængde 10 g
InChI formel InChI=1S/C10H18O8S/c1-10(2)17-8-6(12)7(16-9(8)18-10)5(11)4-15-19(3,13)14/h5-9,11-12H,4H2,1-3H3/t5-,6+,7-,8-,9-/m1/s1
SMIL CC1(C)O[C@H]2O[C@H]([C@H](O)COS(=O)(=O)C)[C@H](O)[C@H]2O1
IUPAC navn [(2R)-2-[(3aR,5R,6S,6aR)-6-hydroxy-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-5-yl]-2-hydroxyethyl] methanesulfonate
Molekylvægt (g/mol) 298.31
Formel vægt 298.0722
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.