missing translation for 'onlineSavingsMsg'
Få mere at vide

1,2’-O-Dimethylguanosine, TRC

Artikelnummer. 30138355
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2.5 mg
10 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30138355 10 mg -
30134434 25 mg -
30003993 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30138355 Leverandør Toronto Research Chemicals Leverandørnr. TRCD46595010MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 73667-71-7
Molekylær formel C12 H17 N5 O5
Molekylvægt (g/mol) 311.294
Synonym 1-Methyl-2'-O-methylguanosine, 2'-O-Methyl-1-methylguanosine
IUPAC navn 2-amino-9-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]-1-methylpurin-6-one
SMIL CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1n2cnc3C(=O)N(C)C(=Nc23)N

Tekniske data

Kemisk navn eller materiale 1,2'-O-Dimethylguanosine
Mængde 10 mg
InChI formel InChI=1S/C12H17N5O5/c1-16-10(20)6-9(15-12(16)13)17(4-14-6)11-8(21-2)7(19)5(3-18)22-11/h4-5,7-8,11,18-19H,3H2,1-2H3,(H2,13,15)/t5-,7-,8-,11-/m1/s1
Formel vægt 311.123
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.