Få mere at vide
1,2-O-Cyclohexylidene myo-Inositol, TRC
Artikelnummer. 30349949
Skift visning
Mængde:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30349949 5 g
30353816 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30349949 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9880005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6763-47-9
Molekylær formel C12 H20 O6
Molekylvægt (g/mol) 260.28
IUPAC navn (3aS,4S,5R,6R,7S,7aR)-spiro[3a,4,5,6,7,7a-hexahydro-1,3-benzodioxole-2,1'-cyclohexane]-4,5,6,7-tetrol
SMIL O[C@H]1[C@@]2([H])OC3(CCCCC3)O[C@@]2([H])[C@@H]([C@H]([C@@H]1O)O)O
Tekniske data
Kemisk navn eller materiale 1,2-O-Cyclohexylidene-myo-inositol
Mængde 5 g
InChI formel InChI=1S/C12H20O6/c13-6-7(14)9(16)11-10(8(6)15)17-12(18-11)4-2-1-3-5-12/h6-11,13-16H,1-5H2/t6-,7-,8+,9+,10-,11+/m1/s1
Formel vægt 260.126