missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Nitrophenyl)piperazine, 95%, Thermo Scientific™

Catalog Number 15403547
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
59084-06-9
C10H14N3O2
208.24
MFCD00040728
YJRCDSXLKPERNV-UHFFFAOYSA-O
1-2-nitrophenyl piperazine, 1-2-nitro-phenyl-piperazine, 2-nitrophenyl piperazine, 1-2-nitrophenyl piperazin, nitrophenylpiperazine, acmc-20ahsu, pubchem14574, 4-nitrophenyl piperazine
100949
1-(2-nitrophenyl)piperazine
[O-][N+](=O)C1=CC=CC=C1N1CC[NH2+]CC1
This item is not returnable. View return policy
59084-06-9
C10H14N3O2
208.24
MFCD00040728
YJRCDSXLKPERNV-UHFFFAOYSA-O
1-2-nitrophenyl piperazine, 1-2-nitro-phenyl-piperazine, 2-nitrophenyl piperazine, 1-2-nitrophenyl piperazin, nitrophenylpiperazine, acmc-20ahsu, pubchem14574, 4-nitrophenyl piperazine
100949
1-(2-nitrophenyl)piperazine
[O-][N+](=O)C1=CC=CC=C1N1CC[NH2+]CC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 207.23
Percent Purity 95%
Chemical Name or Material 1-(2-Nitrophenyl)piperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'