missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Nitrophenyl)homopiperazine, 97%

Brand:  Alfa Aesar™ H63508.03

Additional Details : CAS Number : 450352-64-4 Weight : 0.00100kg Transport : UN number : 3259 Chem class : 8 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15470239

  • 1148.27 DKK / 1g


Estimated Shipment date
from Supplier 20-02-2019
Add to basket

Specifications

Chemical Identifiers

450352-64-4
221.26
1-2-nitrophenyl-1,4-diazepane, 1-2-nitrophenyl-1,4 diazepane, 1h-1,4-diazepine,hexahydro-1-2-nitrophenyl, 1h-1,4-diazepine, hexahydro-1-2-nitrophenyl, 1-2-nitro-phenyl-1,4 diazepane
1-(2-nitrophenyl)-1,4-diazepane
C11H15N3O2
KKGIQPLUVKLSNV-UHFFFAOYSA-N
2829615
C1CNCCN(C1)C2=CC=CC=C2[N+](=O)[O-]

Specifications

1-(2-Nitrophenyl)homopiperazine
450352-64-4
UN3259
KKGIQPLUVKLSNV-UHFFFAOYSA-N
1-(2-nitrophenyl)-1,4-diazepane
2829615
Solid
1g
C11H15N3O2
1-2-nitrophenyl-1,4-diazepane, 1-2-nitrophenyl-1,4 diazepane, 1h-1,4-diazepine,hexahydro-1-2-nitrophenyl, 1h-1,4-diazepine, hexahydro-1-2-nitrophenyl, 1-2-nitro-phenyl-1,4 diazepane
C1CNCCN(C1)C2=CC=CC=C2[N+](=O)[O-]
221.26
221.26
97%
Videos
SDS