Få mere at vide
1-(2-Nitroethyl)-2-naphthol, TRC
Artikelnummer. 30472391
Skift visning
Mængde:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30472391 500 mg
30474646 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30472391 Leverandør Toronto Research Chemicals Leverandørnr. TRCN495895500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 96853-41-7
Molekylær formel C12 H11NO3
Molekylvægt (g/mol) 217.22
IUPAC navn 1-(2-nitroethyl)naphthalen-2-ol
SMIL Oc1ccc2ccccc2c1CC[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale 1-(2-Nitroethyl)-2-naphthol
Mængde 500 mg
InChI formel InChI=1S/C12H11NO3/c14-12-6-5-9-3-1-2-4-10(9)11(12)7-8-13(15)16/h1-6,14H,7-8H2
Formel vægt 217.0739