missing translation for 'onlineSavingsMsg'
Få mere at vide

1,2-Naftalenhydrid, TRC

Product Code. 30472936
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30472936 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30472936 Supplier Toronto Research Chemicals Supplier No. TRCN363003100MG
 Begrænset vare
Forventet forsendelse: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 1,2-Naphthalic Anhydride
CAS 5343-99-7
Molekylær formel C12H6O3
Mængde 100 mg
InChI formel InChI=1S/C12H6O3/c13-11-9-6-5-7-3-1-2-4-8(7)10(9)12(14)15-11/h1-6H
SMIL O=C1OC(=O)c2c1ccc3ccccc23
IUPAC navn benzo[e][2]benzofuran-1,3-Dione
Molekylvægt (g/mol) 198.17
Formel vægt 198.0317
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.