missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[(2-Methyl-4-thiazolyl)methyl]piperazine hydrochloride, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Quantity | 1 g |
Synonym | 2-methyl-4-piperazin-1-ylmethyl thiazole, 1-2-methyl-1,3-thiazol-4-yl methyl piperazine, 1-2-methyl-thiazol-4-ylmethyl-piperazine, 1-2-methyl-thiazol-4-ylmethyl piperazine, 2-methyl-4-piperazin-1-ylmethyl-1,3-thiazole, 2-methyl-4-piperazinylmethyl-1,3-thiazole, 1-2-methylthiazol-4-ylmethyl piperazine, 1-2-methyl-4-thiazolyl methyl piperazine, piperazine,1-2-methyl-4-thiazolyl methyl, piperazine, 1-2-methyl-4-thiazolyl methyl |
Formula Weight | 233.76 |
Chemical Name or Material | 1-[(2-Methyl-4-thiazolyl)methyl]piperazine hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'