Learn More
1-(2-Methoxyphenyl)piperazine Hydrochloride, TRC
Product Code. 30179783
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179783 1 g 1g
30179725 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30179783 Supplier Toronto Research Chemicals Supplier No. TRCM2661851G
 Restricted Item
Forventet forsendelse: 03-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5464-78-8
Molecular Formula C11 H16 N2 O . Cl H
Molecular Weight (g/mol) 228.72
Synonym 1-(2-Methoxyphenyl)piperazine Hydrochloride
IUPAC Name 1-(2-methoxyphenyl)piperazine;hydrochloride
SMILES Cl.COc1ccccc1N2CCNCC2
Specifications
Quantity 1 g
InChI Formula InChI=1S/C11H16N2O.ClH/c1-14-11-5-3-2-4-10(11)13-8-6-12-7-9-13;/h2-5,12H,6-9H2,1H3;1H
Formula Weight 228.1029
Chemical Name or Material 1-(2-Methoxyphenyl)piperazine Hydrochloride