Få mere at vide
1-(2-Methoxy-d3-phenylazo)-2-naphthol, CDN
Artikelnummer. 30312609
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30312609 0.05 g -
30320391 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312609 Leverandør CDN Leverandørnr. CDND73610.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-(2-Methoxyphenylazo)-2-naphthol-D3 (methoxy-D3)
CAS 1398109-09-5
Molekylær formel C17 D3 H11 N2 O2
Mængde 0.05 g
Synonym 1-(2-Methoxy-d3-phenylazo)-2-naphthol, 1-(2-Methoxyphenylazo)-2-naphthol-D3 (methoxy-D3), 1-[2-[2-(Methoxy-d3)phenyl]diazenyl]-2-naphthalenol, Sudan Red G-d3 (methoxy-d3), Anisole-2-azo-β-naphthol-d3
InChI formel InChI=1 S/C17H14N2O2/c1-21-16-9-5-4-8-14(16)18-19-17-13-7-3-2-6-12(13)10-11-15(17)20/h2-11,20 H,1H3/i1D3
SMIL [2 H]C([2 H])([2 H])Oc1ccccc1N=Nc2c(O)ccc3ccccc23
IUPAC navn 1-[[2-(trideuteriomethoxy)phenyl]diazenyl]naphthalen-2-ol
Molekylvægt (g/mol) 281.3238
Formel vægt 281.1244 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity