Learn More
Alfa Aesar™ 1-(2-Isopropylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%
Brand:  Alfa Aesar™ H64079.06
Additional Details : Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15408696
  • 10009.75 DKK / 5g
Estimated Shipment date
from Supplier 07-05-2018
Add to basket
Specifications
Chemical Identifiers
1-(2-Isopropylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
338.32
MFCD26960693
Specifications
1-(2-Isopropylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
Solid
338.32
5g
97%
MFCD26960693