Få mere at vide
1-[2-Hydroxy-3-(1-naphthalenyloxy)propyl]-2,5-pyrrolidinedione-d7, TRC
Artikelnummer. 30404436
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30404436 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30404436 Leverandør Toronto Research Chemicals Leverandørnr. TRCH94860710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-[2-Hydroxy-3-(1-naphthalenyloxy)propyl]-2,5-pyrrolidinedione-d7
CAS 1794942-05-4
Molekylær formel C17H10D7NO4
Mængde 10 mg
InChI formel InChI=1S/C17H17NO4/c19-13(10-18-16(20)8-9-17(18)21)11-22-15-7-3-5-12-4-1-2-6-14(12)15/h1-7,13,19H,8-11H2/i3D,7D,10D2,11D2,13D
SMIL [2H]c1cc2ccccc2c(OC([2H])([2H])C([2H])(O)C([2H])([2H])N3C(=O)CCC3=O)c1[2H]
IUPAC navn 1-[1,1,2,3,3-pentadeuterio-3-(2,3-dideuterionaphthalen-1-yl)oxy-2-hydroxypropyl]pyrrolidine-2,5-dione
Molekylvægt (g/mol) 306.36
Formel vægt 306.1597