missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Fluoro-4-nitrophenyl)-4-piperidone, 97%, Thermo Scientific™

Catalog Number p-7055015
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
439097-58-2
C11H11FN2O3
238.218
BMXFDVAJYBTGRC-UHFFFAOYSA-N
1-2-fluoro-4-nitrophenyl piperidin-4-one, 1-2-fluoro-4-nitrophenyl tetrahydro-4 1h-pyridinone, 1-2-fluoro-4-nitro-phenyl-piperidin-4-one, 4-piperidinone, 1-2-fluoro-4-nitrophenyl, 4-4-oxo-piperidin-1-yl-3-fluoronitrobenzene
3763781
1-(2-fluoro-4-nitrophenyl)piperidin-4-one
C1CN(CCC1=O)C2=C(C=C(C=C2)[N+](=O)[O-])F
This item is not returnable. View return policy
439097-58-2
C11H11FN2O3
238.218
BMXFDVAJYBTGRC-UHFFFAOYSA-N
1-2-fluoro-4-nitrophenyl piperidin-4-one, 1-2-fluoro-4-nitrophenyl tetrahydro-4 1h-pyridinone, 1-2-fluoro-4-nitro-phenyl-piperidin-4-one, 4-piperidinone, 1-2-fluoro-4-nitrophenyl, 4-4-oxo-piperidin-1-yl-3-fluoronitrobenzene
3763781
1-(2-fluoro-4-nitrophenyl)piperidin-4-one
C1CN(CCC1=O)C2=C(C=C(C=C2)[N+](=O)[O-])F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 14265568
Formula Weight 238.22
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(2-Fluoro-4-nitrophenyl)-4-piperidone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'