missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diphenylindole-3-carboxaldehyde, 96%, Thermo Scientific™

Catalog Number 15406087
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
29329-99-5
C21H15NO
297.357
MFCD00629119
VSWUURYDUFEIQQ-UHFFFAOYSA-N
121813
1,2-diphenylindole-3-carbaldehyde
C1=CC=C(C=C1)C2=C(C3=CC=CC=C3N2C4=CC=CC=C4)C=O
This item is not returnable. View return policy
29329-99-5
C21H15NO
297.357
MFCD00629119
VSWUURYDUFEIQQ-UHFFFAOYSA-N
121813
1,2-diphenylindole-3-carbaldehyde
C1=CC=C(C=C1)C2=C(C3=CC=CC=C3N2C4=CC=CC=C4)C=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 229°C to 230°C
Quantity 1 g
Sensitivity Air Sensitive
Synonym 1,2-diphenyl-1h-indole-3-carbaldehyde, 1h-indole-3-carboxaldehyde, 1,2-diphenyl, indole-3-carboxaldehyde, 1,2-diphenyl, 1h-indole-3-carboxaldehyde,1,2-diphenyl, 1,2-diphenylindole-3-carboxaldehyde, 1,2-diphenyl-3-indolecarboxaldehyde
Formula Weight 297.36
Percent Purity 96%
Chemical Name or Material 1,2-Diphenylindole-3-carboxaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'