missing translation for 'onlineSavingsMsg'
Få mere at vide

1,2-Dioleoyl-sn-glycero-3-phospho-rac-(1-glycerol) Sodium Salt, TRC

Artikelnummer. 30372632
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30372632 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30372632 Leverandør Toronto Research Chemicals Leverandørnr. TRCD49362525MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 1,2-Dioleoyl-sn-glycero-3-phospho-rac-(1-glycerol) Sodium Salt
CAS 67254-28-8
Molekylær formel C42H78NaO10P
Mængde 25 mg
Synonym (9Z)-9-Octadecenoic acid (1R)-1-[[[(2,3-dihydroxypropoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl ester, monosodium salt
InChI formel InChI=1S/C42H79O10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-41(45)49-37-40(38-51-53(47,48)50-36-39(44)35-43)52-42(46)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h17-20,39-40,43-44H,3-16,21-38H2,1-2H3,(H,47,48);/q;+1/p-1/b19-17-,20-18-;/t39?,40-;/m1./s1
SMIL O=C(CCCCCCC/C=C\CCCCCCCC)O[C@H](COC(CCCCCCC/C=C\CCCCCCCC)=O)COP(OCC(O)CO)(O[Na])=O
IUPAC navn sodium (R)-2,3-bis(oleoyloxy)propyl (2,3-dihydroxypropyl) phosphate
Molekylvægt (g/mol) 797.03
Formel vægt 796.52
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.