missing translation for 'onlineSavingsMsg'
Få mere at vide

1,2-Dimyristoyl-sn-glycero-3-phosphate Sodium Salt, TRC

Artikelnummer. 30371158
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30371158 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30371158 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4796555MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 1,2-Dimyristoyl-sn-glycero-3-phosphate Sodium Salt
CAS 80724-31-8
Molekylær formel C31H60NaO8P
Mængde 5 mg
Synonym 1,1'-[(1R)-1-[(Phosphonooxy)methyl]-1,2-ethanediyl] Ester Tetradecanoic Acid Sodium Salt, (1R)-1-[(Phosphonooxy)methyl]-1,2-ethanediyl Ester Tetradecanoic Acid Monosodium Salt, 1,2-Dimyristoyl-sn-glycero-3-phosphate Monosodium Salt, Monosodium 1,2-Dimyristoyl-sn-glycerophosphate
InChI formel InChI=1S/C31H61O8P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-30(32)37-27-29(28-38-40(34,35)36)39-31(33)26-24-22-20-18-16-14-12-10-8-6-4-2;/h29H,3-28H2,1-2H3,(H2,34,35,36);/q;+1/p-1/t29-;/m1./s1
SMIL [Na+].CCCCCCCCCCCCCC(=O)OC[C@H](COP(=O)(O)[O-])OC(=O)CCCCCCCCCCCCC
Molekylvægt (g/mol) 614.77
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.