missing translation for 'onlineSavingsMsg'
Få mere at vide

1-(2-Dimethylaminoethyl)-5-mercaptotetrazole, TRC

Artikelnummer. 30377319
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30377319 25 g
30384911 10 g
30371281 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30377319 Leverandør Toronto Research Chemicals Leverandørnr. TRCD46211025G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61607-68-9
Molekylær formel C5H11N5S
Molekylvægt (g/mol) 173.24
Synonym Cefotiam Impurity 17
SMIL CN(C)CCn1nnnc1S

Specifications

Kemisk navn eller materiale 1-(2-Dimethylaminoethyl)-5-mercaptotetrazole
Mængde 25 g
InChI formel InChI=1S/C5H11N5S/c1-9(2)3-4-10-5(11)6-7-8-10/h3-4H2,1-2H3,(H,6,8,11)
Formel vægt 173.0735
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.